Please login to your online account to display your discounted pricing

CD107a (LAMP-1) Rat anti-Mouse, eFluor 450, Clone: eBio1D4B (1D4B), eBioscience™

Rat Monoclonal Antibody

$114.55 - $303.05

Specifications

Antigen CD107a (LAMP-1)
Clone eBio1D4B (1D4B)
Host Species Rat
Gene Alias lysosomal-associated membrane protein 1
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124133

 
life technologies
LS48107180
25μg Each for $114.55

501124134

 
life technologies
LS48107182
100μg Each for $303.05
Description & Specifications

Specifications

Antigen CD107a (LAMP-1)
Clone eBio1D4B (1D4B)
Host Species Rat
Gene Alias lysosomal-associated membrane protein 1
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The eBio1D4B monoclonal antibody reacts with mouse CD107a, also known as lysosomal-associated membrane protein-1 (LAMP-1). CD107a is a type I, lysosomal membrane protein that is extensively glycosylated. It is expressed constitutively in the late endosomes-lysosomes in all cells. CD107a is also transiently expressed on the cell surface of degranulating cytolytic T cells. Additionally, CD107a has been implicated in a variety of cellular functions including cancer metastasis and is also a marker for lysosomal storage disorders.