Please login to your online account to display your discounted pricing

CD107a (LAMP-1) Mouse anti-Human, PE-eFluor 610, Clone: eBioH4A3, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $471.25

Specifications

Antigen CD107a (LAMP-1)
Clone eBioH4A3
Host Species Murine
Gene Alias LAMP1, lysosomal-associated membrane protein 1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124774

 
life technologies
LS61107941
25 Tests Each for $187.05

501124775

 
life technologies
LS61107942
100 Tests Each for $471.25
Description & Specifications

Specifications

Antigen CD107a (LAMP-1)
Clone eBioH4A3
Host Species Murine
Gene Alias LAMP1, lysosomal-associated membrane protein 1
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The eBioH4A3 monoclonal antibody reacts with human CD107a, also known as lysosomal-associated membrane protein-1 (LAMP-1). CD107a is a highly glycosylated protein of approximately 110kDa. It is predominantly expressed intracellularly in the lysosomal/endosomal membrane in nearly all cells. CD107a is transiently expressed on the cell surface of degranulating cytolytic T cells, and is also upregulated on the surface of activated platelets and some cancer cells.