missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD107a (LAMP-1) Mouse anti-Human, PE-Cy5, Clone: eBioH4A3, eBioscience™

Mouse Monoclonal Antibody

$117.48 - $348.48

Specifications

Antigen CD107a (LAMP-1)
Clone eBioH4A3
Host Species Murine
Gene Alias LAMP1, lysosomal-associated membrane protein 1
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014088 Life Technologies
LS15107941
25 Tests Each for $117.48
5014089 Life Technologies
LS15107942
100 Tests Each for $348.48
Specifications

Description

The eBioH4A3 monoclonal antibody reacts with human CD107a, also known as lysosomal-associated membrane protein-1 (LAMP-1). CD107a is a highly glycosylated protein of approximately 110kDa. It is predominantly expressed intracellularly in the lysosomal/endosomal membrane in nearly all cells. CD107a is transiently expressed on the cell surface of degranulating cytolytic T cells, and is also upregulated on the surface of activated platelets and some cancer cells.
Description & Specifications

Specifications

CD107a (LAMP-1)
Murine
Flow Cytometry (Intracellular Staining)
PE-Cy5
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test
eBioH4A3
LAMP1, lysosomal-associated membrane protein 1
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer