missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD106 (VCAM-1) Rat anti-Mouse, eFluor 450, Clone: 429, eBioscience™

Rat Monoclonal Antibody

$201.55 - $404.55

Specifications

Antigen CD106 (VCAM-1)
Clone 429
Host Species Rat
Gene Alias VCAM1, vascular cell adhesion molecule-1
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124132 Life Technologies
LS48106180
25μg Each for $201.55
5016323 Life Technologies
LS48106182
100μg Each for $404.55
Specifications

Description

The 429 monoclonal antibody reacts with mouse CD106 (Vascular Cell Adhesion Molecule-1, VCAM-1), a 110kDa transmembrane glycoprotein expressed by myeloid lineage and bone marrow stromal cells. Endothelial cells constitutively express low levels of CD106 and upregulate it upon cytokine stimulation. CD106 binds to integrin α4β1 (VLA-4, CD49d/CD29) and Integrin a4b7 (LPAM-1) and these interactions in the bone marrow and thymus are important for early lymphocyte and myeloid development. Cytokine-mediated upregulation of CD106 on endothelial cells suggests a role for this antigen in the inflammatory response.
Description & Specifications

Specifications

CD106 (VCAM-1)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
429
VCAM1, vascular cell adhesion molecule-1
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL