missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD106 (VCAM-1) Rat anti-Mouse, Clone: 429, eBioscience™

Rat Monoclonal Antibody

$77.88 - $289.08

Specifications

Antigen CD106 (VCAM-1)
Clone 429
Host Species Rat
Gene Alias VCAM1, vascular cell adhesion molecule-1
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012805 Life Technologies
LS14106181
50μg Each for $77.88
5012806 Life Technologies
LS14106182
100μg Each for $130.68
5012807 Life Technologies
LS14106185
500μg Each for $289.08
Specifications

Description

The 429 monoclonal antibody reacts with mouse CD106 (Vascular Cell Adhesion Molecule-1, VCAM-1), a 110kDa transmembrane glycoprotein expressed by myeloid lineage and bone marrow stromal cells. Endothelial cells constitutively express low levels of CD106 and upregulate it upon cytokine stimulation. CD106 binds to integrin α4β1 (VLA-4, CD49d/CD29) and Integrin a4b7 (LPAM-1) and these interactions in the bone marrow and thymus are important for early lymphocyte and myeloid development. Cytokine-mediated upregulation of CD106 on endothelial cells suggests a role for this antigen in the inflammatory response.
Description & Specifications

Specifications

CD106 (VCAM-1)
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
429
VCAM1, vascular cell adhesion molecule-1
Flow Cytometry,Functional Assay,Immunohistochemistry (Frozen),Immunoprecipitation
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL