missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD106 (VCAM-1) Mouse anti-Rat, Biotin, Clone: eBioMR106 (MR106), eBioscience™

Mouse Monoclonal Antibody

$130.68 - $209.88

Specifications

Antigen CD106 (VCAM-1)
Clone eBioMR106 (MR106)
Host Species Murine
Gene Alias VCAM1, vascular cell adhesion molecule-1
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011694 Life Technologies
LS13106081
50μg Each for $130.68
5011695 Life Technologies
LS13106082
100μg Each for $209.88
Specifications

Description

The eBioMR106 monoclonal antibody reacts with rat CD106 (Vascular Cell Adhesion Molecule-1, VCAM-1), a 110kDa transmembrane glycoprotein expressed by myeloid lineage and bone marrow stromal cells. Endothelial cells constitutively express low levels of CD106 and further upregulate VCAM-1 upon cytokine stimulation. CD106 binds to integrin α4β1 (VLA-4, CD49d/CD29) and Integrin α4β7 (LPAM-1) and these interactions in the bone marrow and thymus are important for early lymphocyte and myeloid development. Cytokine-mediated upregulation of CD106 on endothelial cells suggests a role for this antigen in the inflammatory response.
Description & Specifications

Specifications

CD106 (VCAM-1)
Murine
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
eBioMR106 (MR106)
VCAM1, vascular cell adhesion molecule-1
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer