Learn More
CD106 (VCAM-1) Monoclonal Antibody (STA), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen 12106941
Description
Description: The STA monoclonal antibody reacts with human CD106 (Vascular Cell Adhesion Molecule-1, VCAM-1), a 110 kDa transmembrane glycoprotein expressed by myeloid lineage and bone marrow stromal cells. Endothelial cells constitutively express low levels of CD106 and upregulate it upon cytokine stimulation. CD106 binds to Integrin alpha4beta1 (VLA-4, CD49d/CD29) and Integrin alpha4beta7 (LPAM-1). Cytokine-mediated upregulation of CD106 on endothelial cells suggests a role for this antigen in the inflammatory response. Applications Reported: STA has been reported for use in flow cytometric analysis. Applications Tested: This STA antibody has been pre-titrated and tested by flow cytometric analysis of stimulated HUVEC. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.Specifications
CD106 (VCAM-1) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P19320 | |
VCAM1 | |
Affinity chromatography | |
RUO | |
7412 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
STA | |
PE | |
VCAM1 | |
adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.