Please login to your online account to display your discounted pricing

CD106 (VCAM-1) Mouse anti-Human, FITC, Clone: STA, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen CD106 (VCAM-1)
Clone STA
Host Species Murine
Gene Alias vascular cell adhesion molecule-1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122086

 
life technologies
LS11106941
25 Tests Each for $143.55

501122087

 
life technologies
LS11106942
100 Tests Each for $361.05
Description & Specifications

Specifications

Antigen CD106 (VCAM-1)
Clone STA
Host Species Murine
Gene Alias vascular cell adhesion molecule-1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (1μg)/test

The STA monoclonal antibody reacts with human CD106 (Vascular Cell Adhesion Molecule-1, VCAM-1), a 110kDa transmembrane glycoprotein expressed by myeloid lineage and bone marrow stromal cells. Endothelial cells constitutively express low levels of CD106 and upregulate it upon cytokine stimulation. CD106 binds to Integrin α4β1 (VLA-4, CD49d/CD29) and Integrin α4β7 (LPAM-1). Cytokine-mediated upregulation of CD106 on endothelial cells suggests a role for this antigen in the inflammatory response.