missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD106 (VCAM-1) Mouse anti-Human, Biotin, Clone: STA, eBioscience™

Mouse Monoclonal Antibody

$104.28 - $236.28

Specifications

Antigen CD106 (VCAM-1)
Clone STA
Host Species Murine
Gene Alias vascular cell adhesion molecule-1
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011700 Life Technologies
LS13106980
25μg Each for $104.28
5011701 Life Technologies
LS13106982
100μg Each for $236.28
Specifications

Description

The STA monoclonal antibody reacts with human CD106 (Vascular Cell Adhesion Molecule-1, VCAM-1), a 110kDa transmembrane glycoprotein expressed by myeloid lineage and bone marrow stromal cells. Endothelial cells constitutively express low levels of CD106 and upregulate it upon cytokine stimulation. CD106 binds to Integrin α4β1 (VLA-4, CD49d/CD29) and Integrin α4β7 (LPAM-1). Cytokine-mediated upregulation of CD106 on endothelial cells suggests a role for this antigen in the inflammatory response.
Description & Specifications

Specifications

CD106 (VCAM-1)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
STA
vascular cell adhesion molecule-1
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL