Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD106/VCAM-1 Ab-3, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ CD106/VCAM-1 Ab-3, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1101PABX

 View more versions of this product

Catalog No. MS1101PABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Immunohistochemistry (Paraffin)
Isotype IgG1
Quantity 200µL
Antigen CD106/VCAM-1 Ab-3
Immunogen Stimulated human umbilical vein endothelial cells (HUVEC)
Cross Reactivity Human
Format Concentrated
Regulatory Status RUO
Clone 1.4C3
Research Discipline Inflammation
Formulation Ab without BSA/Azide (Frozen)
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled
Item Description 200uL; 1mg/mL; Purified w/o BSA, azide

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ CD106/VCAM-1 Ab-3, Mouse Monoclonal Antibody.

 

CD106 (also known as vascular cell adhesion molecule-1 [VCAM-1] and INCAM-110) is a member of the Ig superfamily of adhesion molecules and is expressed at high levels on cytokine-stimulated vascular endothelial cells, and at minimal levels on unstimulated endothelial cells. It is also present on follicular and interfollicular dendritic cells of lymph nodes, myoblasts, and some macrophages. CD106 serves as a ligand for leukocyte integrin alphaBeta (VLA-4 or CD49d/CD29) and mediates cell adhesion of leukocytes to activated endothelium. It plays a role in tumor metastasis.

Host Species: Mouse

Clone: 1.4C3

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Stimulated human umbilical vein endothelial cells (HUVEC)

Molecular Weight: 110 kDa

Positive Control: Placenta

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen and formalin/paraffin)
  • Western Blotting (Not recommended)

Note:

USA: RUO; Int'l: RUO