Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD105/Endoglin/TGF-β 1/3 Receptor Ab-3, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting and flow cytometry applications.

Manufacturer: thermo scientific™ lab vision™  MS1290PABX

 View more versions of this product

Catalog No. MS1290PABX

  • / Each
 

Description & Specifications

Specifications

Immunogen Purified GP160 from cell membrane glycoproteins of fresh non-T/non-B acute lymphoblastic leukemia cells
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Isotype IgG2a kappa
Quantity 200µL
Antigen CD105/Endoglin/TGF-beta1/3 Receptor Ab-3
Clone SN6h
Regulatory Status RUO
Type Primary
Classification Monoclonal
Conjugate Unlabeled
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Angiogenesis
Concentration 1mg/mL
Item Description 200uL; 1mg/mL; Purified w/o BSA, azide

Ensure accurate, reproducible results in immunohistochemistry, western blotting and flow cytometry applications with Thermo Scientific™ CD105/Endoglin/TGF-β1/3 Receptor Ab-3, Mouse Monoclonal Antibody.

CD105 or Endoglin or GP160 is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists as an O- and N-glycosylated homodimer. Up-regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation-associated endothelial cell marker. CD105 binds to TGF-Beta1 and Beta3 with high affinity but not TGF-Beta2.

Host Species: Mouse

Clone: SN6h

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Purified GP160 from cell membrane glycoproteins of fresh non-T/non-B acute lymphoblastic leukemia cells.

Molecular Weight: 95 kDa (monomer) and 190kDa (dimer)

Positive Control: HUVEC cells, Tonsil, lymph node and thymus

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO