missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD105 (Endoglin) Rat anti-Mouse, PE-Cy7, Clone: MJ7/18, eBioscience™

Rat Monoclonal Antibody

$174.24 - $443.52

Specifications

Antigen CD105 (Endoglin)
Clone MJ7/18
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123318 Life Technologies
LS25105180
25μg Each for $174.24
501123319 Life Technologies
LS25105182
100μg Each for $443.52
Specifications

Description

The MJ7/18 monoclonal antibody reacts with the mouse CD105 molecule, also known as Endoglin. This 90kDa disulfide-linked homodimer is expressed by vascular endothelial cells. It is suggested that CD105 functions in adhesion and embryonic angiogenesis.
Description & Specifications

Specifications

CD105 (Endoglin)
Rat
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL
MJ7/18
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer