missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD105 (Endoglin) Rat anti-Mouse, Clone: MJ7/18, eBioscience™

Rat Monoclonal Antibody

$91.08 - $328.68

Specifications

Antigen CD105 (Endoglin)
Clone MJ7/18
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012797 Life Technologies
LS14105181
50μg Each for $91.08
5012798 Life Technologies
LS14105182
100μg Each for $130.68
5012799 Life Technologies
LS14105185
500μg Each for $328.68
Specifications

Description

The MJ7/18 monoclonal antibody reacts with the mouse CD105 molecule, also known as Endoglin. This 90kDa disulfide-linked homodimer is expressed by vascular endothelial cells. It is suggested that CD105 functions in adhesion and embryonic angiogenesis.
Description & Specifications

Specifications

CD105 (Endoglin)
Rat
Flow Cytometry,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
MJ7/18
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer