missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD105 (Endoglin) Rat anti-Mouse, Biotin, Clone: MJ7/18, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS13105181

 View more versions of this product

Catalog No. 501128897



Description

Description

Description: The MJ7/18 monoclonal antibody reacts with the mouse CD105 molecule, also known as Endoglin. This 90 kDa disulfide-linked homodimer is expressed by vascular endothelial cells. It is suggested that CD105 functions in adhesion and embryonic angiogenesis. Applications Reported: The MJ7/18 antibody has been reported for use in flow cytometric analysis. Applications Tested: The MJ7/18 antibody has been tested by flow cytometric analysis of bEnd.3 cells and mouse splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.
Specifications

Specifications

CD105 (Endoglin)
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q63961
Endoglin
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
MJ7/18
Biotin
Eng
Liquid
Eng
IgG2a, kappa
50 μg
4° C, store in dark, DO NOT FREEZE!
Primary
13805
Documents

For Research Use Only.