missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD105 (Endoglin) Rat anti-Mouse, Biotin, Clone: MJ7/18, eBioscience™

Rat Monoclonal Antibody

$114.55 - $390.05

Specifications

Antigen CD105 (Endoglin)
Clone MJ7/18
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122519 Life Technologies
LS13105185
500μg Each for $390.05
5011692 Life Technologies
LS13105182
100μg Each for $288.55
501128897 Life Technologies
LS13105181
50μg Each for $114.55
Specifications

Description

The MJ7/18 monoclonal antibody reacts with the mouse CD105 molecule, also known as Endoglin. This 90kDa disulfide-linked homodimer is expressed by vascular endothelial cells. It is suggested that CD105 functions in adhesion and embryonic angiogenesis.
Description & Specifications

Specifications

CD105 (Endoglin)
Rat
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
MJ7/18
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer