Please login to your online account to display your discounted pricing

CD105 (Endoglin) Mouse anti-Human, PE, Clone: SN6, eBioscience™

Mouse Monoclonal Antibody

$147.90 - $321.90

Specifications

Antigen CD105 (Endoglin)
Clone SN6
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010551

 
life technologies
LS12105741
25 Tests Each for $147.90

5010552

 
life technologies
LS12105742
100 Tests Each for $321.90
Description & Specifications

Specifications

Antigen CD105 (Endoglin)
Clone SN6
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (1μg)/test (a test is defined as the amount that will stain 1 x 10e6 cells in 100μL)

The SN6 monoclonal antibody reacts with human CD105, also known as Endoglin. CD105, an approximately 90kDa disulfide-linked homodimer is expressed by vascular endothelial cells and some bone marrow cells and activated macrophages. It is suggested that CD105 functions in adhesion and embryonic angiogenesis.