missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD104 (Integrin beta 4) Rat anti-Human, Clone: 439-9B, eBioscience™

Rat Monoclonal Antibody

$77.88 - $143.88

Specifications

Antigen CD104 (Integrin beta 4)
Clone 439-9B
Host Species Rat
Gene Alias Integrin b4, ITGB4
Applications Flow Cytometry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012795 Life Technologies
LS14104980
25μg Each for $77.88
5012796 Life Technologies
LS14104982
100μg Each for $143.88
Specifications

Description

The monoclonal antibody 439-9B recognizes human CD104, also known as integrin beta 4. CD104 is a 202kDa subunit that associates with integrin alpha 6 (CD49f), thereby forming the laminin receptor. Multiple splice variants have been identified. Expression is found on epithelial cells and tumor cells and has been shown to be involved in cell-matrix and cell-cell interactions. The role of CD104 in tumorigenicity is well established. The monoclonal antibody 439-9B has been reported to have no effect on adhesion of human colon adenocarcinoma cells. Moreover, this antibody crossreacts to bovine.
Description & Specifications

Specifications

CD104 (Integrin beta 4)
Rat
Flow Cytometry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
439-9B
Integrin b4, ITGB4
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer