Please login to your online account to display your discounted pricing

CD103 (Integrin alpha E) Mouse anti-Rat, PE, Clone: OX62, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $274.05

Specifications

Antigen CD103 (Integrin alpha E)
Clone OX62
Host Species Murine
Gene Alias Integrin aE, ITGAE
Species Reactivity Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010539

 
life technologies
LS12103080
25μg Each for $129.05

5010540

 
life technologies
LS12103082
100μg Each for $274.05
Description & Specifications

Specifications

Antigen CD103 (Integrin alpha E)
Clone OX62
Host Species Murine
Gene Alias Integrin aE, ITGAE
Species Reactivity Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The OX62 monoclonal antibody reacts with rat CD103, also known as αE2 integrin. CD103 was originally identified on rat veiled dendritic cells, and is expressed by dendritic epidermal T cells in the skin, intraepithelial lymphocytes in the small intestine and by dendritic cells associated with γδ T cells. Integrins are a family of heterodimeric proteins which contain non-covalently associated α and β transmembrane glycoproteins. Integrins function as receptors for extracellular matrix and cell-cell adhesion molecules.