Please login to your online account to display your discounted pricing

CD103 (Integrin alpha E) Mouse anti-Human, PerCP-eFluor 710, Clone: Ber-ACT8, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $375.55

Specifications

Antigen CD103 (Integrin alpha E)
Clone Ber-ACT8
Host Species Mouse
Gene Alias Integrin alpha-E, ITGAE
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501129424

 
life technologies
LS46103741
25 tests Each for $172.55

501130774

 
life technologies
LS46103742
100 tests Each for $375.55
Description & Specifications

Specifications

Antigen CD103 (Integrin alpha E)
Clone Ber-ACT8
Host Species Mouse
Gene Alias Integrin alpha-E, ITGAE
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

The Ber-ACT8 monoclonal antibody reacts with human CD103, the alpha E integrin. CD103 non-covalently associates with integrin beta7. CD103 is expressed mainly on intraepithelial lymphocytes and a small subset of peripheral lymphocytes. CD103 is also expressed by hairy cell leukemia (HCL) and by some chronic B cell lymphocytic leukemias. In vitro stimulation of human T cells with mitogens induces upregulation of CD103. Epithelial cell antigen (E-cadherin) binds to CD103 and mediates homing of lymphocytes to the intestinal epithelium. Cross-blocking studies suggest that B-Ly7 and Ber-ACT8 see different epitopes.