missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD103 (Integrin alpha E) Mouse anti-Human, PE-Cy7, Clone: B-Ly7, eBioscience™

Mouse Monoclonal Antibody

$196.68 - $394.68

Specifications

Antigen CD103 (Integrin alpha E)
Clone B-Ly7
Host Species Murine
Gene Alias ITGAE
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015540 Life Technologies
LS25103841
25 Tests Each for $196.68
5015541 Life Technologies
LS25103842
100 Tests Each for $394.68
Specifications

Description

The B-Ly7 monoclonal antibody reacts with human CD103, the alpha E integrin. CD103 non-covalently associates with integrin beta 7. CD103 is expressed mainly on intraepithelial lymphocytes and a small subset of peripheral lymphocytes. CD103 is also expressed by hairy cell leukemia (HCL) and by some chronic B cell lymphocytic leukemias. In vitro stimulation of human T cells with mitogens induces upregulation of CD103. Epithelial cell antigen, E-cadherin, binds to CD103 and mediates homing of lymphocytes to the intestinal epithelium.
Description & Specifications

Specifications

CD103 (Integrin alpha E)
Murine
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.25μg)/test
B-Ly7
ITGAE
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer