Please login to your online account to display your discounted pricing

CD103 (Integrin alpha E) Mouse anti-Human, APC, Clone: B-Ly7, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $361.05

Specifications

Antigen CD103 (Integrin alpha E)
Clone B-Ly7
Host Species Mouse
Gene Alias ITGAE
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015045

 
life technologies
LS17103841
25 tests Each for $158.05

5015046

 
life technologies
LS17103842
100 tests Each for $361.05
Description & Specifications

Specifications

Antigen CD103 (Integrin alpha E)
Clone B-Ly7
Host Species Mouse
Gene Alias ITGAE
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The B-Ly7 monoclonal antibody reacts with human CD103, the alpha E integrin. CD103 non-covalently associates with integrin beta 7. CD103 is expressed mainly on intraepithelial lymphocytes and a small subset of peripheral lymphocytes. CD103 is also expressed by hairy cell leukemia (HCL) and by some chronic B cell lymphocytic leukemias. In vitro stimulation of human T cells with mitogens induces upregulation of CD103. Epithelial cell antigen, E-cadherin, binds to CD103 and mediates homing of lymphocytes to the intestinal epithelium.