Please login to your online account to display your discounted pricing

CD102 (ICAM-2) Rat anti-Mouse, eFluor 450, Clone: 3C4 (mIC2/4), eBioscience™

Rat Monoclonal Antibody

$158.05 - $332.05

Specifications

Antigen CD102 (ICAM-2)
Clone 3C4 (mIC2/4)
Host Species Rat
Gene Alias ICAM2, Ly-60
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016318

 
life technologies
LS48102180
25μg Each for $158.05

5016319

 
life technologies
LS48102182
100μg Each for $332.05
Description & Specifications

Specifications

Antigen CD102 (ICAM-2)
Clone 3C4 (mIC2/4)
Host Species Rat
Gene Alias ICAM2, Ly-60
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody 3C4 (mIC2/4) recognizes mouse CD102, also known as intercellular adhesion molecule-2 (ICAM-2). CD102, a member of the Immunoglobulin superfamily of receptors with 2 extracellular Ig domains, binds LFA (CD11a/CD18) and Mac-1 (CD11b/CD18), thereby mediating cellular interactions to lymphocytes. CD102 plays a role in NK cytotoxicity and migration, and lymphocyte recirculation. Expression of CD102 is found on most lymphocytes, monocytes and platelets.

The 3C4 (mIC2/4) antibody blocks interaction of lymphocytes with antigen presenting cells.