Please login to your online account to display your discounted pricing

CD102 (ICAM-2) Rat anti-Mouse, Clone: 3C4 (mIC2/4), Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$159.50 - $406.00

Specifications

Antigen CD102 (ICAM-2)
Clone 3C4 (mIC2/4)
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122945

 
life technologies
LS16102185
500μg Each for $406.00

501122944

 
life technologies
LS16102182
100μg Each for $159.50
Description & Specifications

Specifications

Antigen CD102 (ICAM-2)
Clone 3C4 (mIC2/4)
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The monoclonal antibody 3C4 (mIC2/4) recognizes mouse CD102, also known as intercellular adhesion molecule-2 (ICAM-2). CD102, a member of the Immunoglobulin superfamily of receptors with 2 extracellular Ig domains, binds LFA (CD11a/CD18) and Mac-1 (CD11b/CD18), thereby mediating cellular interactions to lymphocytes. CD102 plays a role in NK cytotoxicity and migration, and lymphocyte recirculation. Expression of CD102 is found on most lymphocytes, monocytes and platelets.

The 3C4 (mIC2/4) antibody blocks interaction of lymphocytes with antigen presenting cells.