missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD102 (ICAM-2) Mouse anti-Human, Clone: CBRIC2/2, eBioscience™

Mouse Monoclonal Antibody

$85.55 - $143.88

Specifications

Antigen CD102 (ICAM-2)
Clone CBRIC2/2
Host Species Murine
Gene Alias ICAM2
Applications Flow Cytometry,Functional Assay,Immunohistochemistry,Immunoprecipitation,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012788 Life Technologies
LS14102980
25μg Each for $85.55
5012789 Life Technologies
LS14102982
100μg Each for $143.88
Specifications

Description

The CBRIC2/2 monoclonal antibody reacts with human ICAM-2 (CD102). ICAM-2 is a 55-65 kD transmembrane glycoprotein possessing 2 extracellular Ig domains, a single transmembrane domain, and a short 26-amino acid cytoplasmic domain. ICAM-2 is expressed on most leukocytes, and is strongly expressed on vascular endothelial cells. ICAM-2 is a ligand of integrin CD11a/CD18 (LFA-1), and also has been reported to bind integrin CD11b/CD18 (Mac-1). ICAM-2 is an intercellular adhesion molecule involved in lymphocyte homing to sites of inflammation. The CBRIC2/2 antibody has been reported to have blocking activity.
Description & Specifications

Specifications

CD102 (ICAM-2)
Murine
Flow Cytometry,Functional Assay,Immunohistochemistry,Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
CBRIC2/2
ICAM2
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer