Please login to your online account to display your discounted pricing

CD100 (SEMA4D) Rat anti-Mouse, FITC, Clone: BMA12 (BMA-12), eBioscience™

Rat Monoclonal Antibody

$172.55 - $303.05

Specifications

Antigen CD100 (SEMA4D)
Clone BMA12 (BMA-12)
Host Species Rat
Gene Alias Semaphorin 4D
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509718

 
affymetrix
11100181
50μg Each for $172.55

509719

 
affymetrix
11100182
100μg Each for $303.05
Description & Specifications

Specifications

Antigen CD100 (SEMA4D)
Clone BMA12 (BMA-12)
Host Species Rat
Gene Alias Semaphorin 4D
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The BMA12 monoclonal antibody reacts with mouse CD100, also known as Sema4D. Sema4D/CD100 is a 150kDa transmembrane protein expressed constitutively on T cells and is involved in the activation of B cells through its interaction with CD72. Interaction of these two proteins diminishes the negative signaling normally effected by CD72. CD100 is already expressed strongly on resting T cells and somewhat weakly on B cells, but activation upregulates expression on both cell types. Antigen expression has been reported in Spleen, thymus and lymph nodes.