missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD10 Mouse anti-Human, PerCP-eFluor 710, Clone: SN5c, eBioscience™

Mouse Monoclonal Antibody

$473.88 - $473.88

Specifications

Antigen CD10
Clone SN5c
Host Species Murine
Gene Alias Common Acute Lymphocytic Leukemia antigen (CALLA), Neprilysin
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015914 Life Technologies
LS46010842
100 Tests Each for $473.88
Specifications

Description

The eBioSN5c monoclonal antibody recognizes human CD10 (CALLA, NEP, enkephalinase, Neprilysin), which is a 100 kDa, type II cell surface glycoprotein originally identified for its expression on most acute lymphoblastic leukemias (ALLs). Subsequently, CD10 was shown to be the same molecule as neutral endopeptidase (NEP), or KII-NA. CD10 is a Zn2+-dependent metallopeptidase acting on substrates such as endothelin, glucagon, gastrin, neurotensin and bradykinin. CD10 is involved in the regulation of chemotactic and inflammatory processes involving neutrophils. In B cells, CD10 regulates stromal cell-dependent B lymphopoiesis and its expression has been reported on mature germinal center B cells. CD10 expression is also found on normal donor granulocytes and bone marrow stromal cells. Most likely, CB-CALLA and eBioSN5c see different epitopes due to their ability to co-stain.
Description & Specifications

Specifications

CD10
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
SN5c
Common Acute Lymphocytic Leukemia antigen (CALLA), Neprilysin
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test