Please login to your online account to display your discounted pricing

CD10 Mouse anti-Human, Alexa Fluor 700, Clone: CB-CALLA, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $419.05

Specifications

Antigen CD10
Clone eBioCB-CALLA (CB-CALLA)
Host Species Murine
Gene Alias Common Acute Lymphocytic Leukemia antigen (CALLA), Neprilysin
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124671

 
life technologies
LS56010641
25 Tests Each for $187.05

501124672

 
life technologies
LS56010642
100 Tests Each for $419.05
Description & Specifications

Specifications

Antigen CD10
Clone eBioCB-CALLA (CB-CALLA)
Host Species Murine
Gene Alias Common Acute Lymphocytic Leukemia antigen (CALLA), Neprilysin
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Alexa Fluor 700
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The eBioCB-CALLA monoclonal antibody recognizes human CD10 (CALLA, NEP, enkephalinase, Neprilysin), which is a 100 kDa, type II cell surface glycoprotein originally identified for its expression on most acute lymphoblastic leukemias (ALL). Subsequently, CD10 was shown to be the same molecule as the neutral endopeptidase (NEP), or KII-NA. CD10 is a Zn2+-dependent metallo-peptidase with endothelin, glucagon, gastrin, neurotensin and bradykinin included among its substrates. CD10 is involved in the regulation of chemotactic and inflammatory processes involving neutrophils. In B cells, CD10 regulates stromal cell-dependent B lymphopoiesis and expression has also been reported on mature B cells in germinal centres. In addition to the hematopoietic compartment, other major sites of CD10 expression are the brush border of enterocytes and renal tubules and glomeruli. There is partial blocking of the eBioCB-CALLA and MEM-78 monoclonal antibodies indicating that they recognize similar epitopes.