Please login to your online account to display your discounted pricing

CD 2665, Tocris™

Selective RARβ/γ antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material CD 2665
Assay Percent Range >98%
Molecular Formula C31H34O5
Formula Weight 486.6
Synonym 4-[6-[(2-Methoxyethoxy)methoxy]-7-tricyclo[3.3.1.13, 7]dec-1-yl-2-naphthalenyl)benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

380010

 
r & d systems
RDS380010
10mg Each for $252.39

380050

 
r & d systems
RDS380050
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 486.6
Quantity 10mg
CAS 170355-78-9
Assay Percent Range >98%
Molecular Formula C31H34O5
Recommended Storage Store at +4°C
Synonym 4-[6-[(2-Methoxyethoxy)methoxy]-7-tricyclo[3.3.1.13, 7]dec-1-yl-2-naphthalenyl)benzoic acid
Chemical Name or Material CD 2665
Safety and Handling

Store at +4°C