Please login to your online account to display your discounted pricing

CCT 031374 hydrobromide, Tocris™

Inhibits TCF-dependent transcription; lowers β-catenin levels

$208.25 - $867.95

Chemical Identifiers

Chemical Name or Material CCT 031374 hydrobromide
Assay Percent Range >98%
Molecular Formula C23H19N3O·HBr
Formula Weight 434.33
Synonym 1-[1,1'-Biphenyl]-4-yl-2-(2,3-dihydro-9H-imidazo[1,2-a]benzimidazol-9-yl)ethanone hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

467550

 
r & d systems
467550
50mg Each for $867.95

467510

 
r & d systems
467510
10mg Each for $208.25
Description & Specifications

Specifications

Formula Weight 434.33
Quantity 50mg
Chemical Name or Material CCT 031374 hydrobromide
Synonym 1-[1,1'-Biphenyl]-4-yl-2-(2,3-dihydro-9H-imidazo[1,2-a]benzimidazol-9-yl)ethanone hydrobromide
Assay Percent Range >98%
CAS 1219184-91-4
Molecular Formula C23H19N3O·HBr
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature