Please login to your online account to display your discounted pricing

CCMQ, Tocris


Used to characterize NR2B-containing NMDA receptors

Quantity 10mg

Manufacturer: r & d systems  RDS123810

 View more versions of this product

Catalog No. 123810

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 231.21
Quantity 10mg
Chemical Name or Material CCMQ
Assay Percent Range >98%
CAS 132623-44-0
Molecular Formula C12H9NO4
Synonym 2-Carboxy-3-carboxymethylquinoline
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material CCMQ
Assay Percent Range >98%
Molecular Formula C12H9NO4
Formula Weight 231.21
Synonym 2-Carboxy-3-carboxymethylquinoline
Accessories

Desiccate at +4°C