Please login to your online account to display your discounted pricing

CCMQ, Tocris

Used to characterize NR2B-containing NMDA receptors

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material CCMQ
Assay Percent Range >98%
Molecular Formula C12H9NO4
Formula Weight 231.21
Synonym 2-Carboxy-3-carboxymethylquinoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

123810

 
r & d systems
RDS123810
10mg Each for $177.37

123850

 
r & d systems
RDS123850
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 231.21
Quantity 10mg
Chemical Name or Material CCMQ
Assay Percent Range >98%
CAS 132623-44-0
Molecular Formula C12H9NO4
Synonym 2-Carboxy-3-carboxymethylquinoline
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C