Learn More
CCL8 (MCP-2) Mouse anti-Human, eFluor™ 660, Clone: DWZEE, eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen 50978941
Description
Description: This DWZEE monoclonal antibody reacts with human CCL8. CCL8, also known as MCP-2 (Monocyte Chemotactic Protein 2), is a member of the CC- subfamily of chemokines and is most closely related to CCL2 (MCP-1) and CCL7 (MCP-3). They are secreted by a variety of cell types in response to inflammatory stimuli and play critical roles in the recruitment of leukocytes to areas of inflammation. While all three MCP proteins are potent chemoattractants of monocytes and T cells, CCL7 appears to have the broadest range of activity, as it has also been demonstrated to attract activated NK cells, eosinophils, basophils, and neutrophils. CCL8 has also been demonstrated to attract NK cells, eosinophils, and basophils, but requires higher concentrations to do so. CCL8 signals via the g protein-coupled receptor CCR5, which is shared with other CC-chemokines. CCR5 is the primary co-receptor for HIV entry, which the virus binds through the gp120 envelope protein. All CCR5 ligands demonstrate potent inhibition of virus entry into the cell, both through steric hindrance of gp120-CCR5 interaction, and ligand-mediated receptor internalization. Applications Reported: This DWZEE antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This DWZEE antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood monocytes.
This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes, lymphocytes, basophils and eosinophils. By recruiting leukocytes to sites of inflammation this cytokine may contribute to tumor-associated leukocyte infiltration and to the antiviral state against HIV infection.Specifications
CCL8 (MCP-2) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P80075 | |
CCL8 | |
Affinity chromatography | |
RUO | |
6355 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
DWZEE | |
eFluor 660 | |
CCL8 | |
HC14, MCP-2, MCP2, SCYA10, SCYA8 | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.