Please login to your online account to display your discounted pricing

CCL2 (MCP-1) Mouse anti-Human, PE, Clone: 5D3-F7, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $143.55

Specifications

Antigen CCL2 (MCP-1)
Clone 5D3-F7
Host Species Mouse
Gene Alias MCAF
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122376

 
life technologies
LS12709980
25μg Each for $143.55
Description & Specifications

Specifications

Antigen CCL2 (MCP-1)
Clone 5D3-F7
Host Species Mouse
Gene Alias MCAF
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 5D3-F7 antibody reacts with human, cynomolgus, and rhesus monkey monocyte chemoattractant protein-1 (MCP-1), also known as monocyte chemotactic and activating factor (MCAF) and CCL2. MCP-1 is a CC chemokine which presents as a dimer. Derived from monocytes and vascular endothelial cells, it exerts chemoattractant activity on a variety of cell types, such as monocytes, basophils, T cells and NK cells via chemokine receptors such as CCR2, CCR5, and CCR10.