Please login to your online account to display your discounted pricing

CCL2 (MCP-1) Armenian Hamster, Biotin, Clone: 2H5, eBioscience™

Armenian Hamster Monoclonal Antibody

$158.05 - $404.55

Specifications

Antigen CCL2 (MCP-1)
Clone 2H5
Host Species Armenian Hamster
Gene Alias MCP1, MCAF, CCL-2
Species Reactivity Human, Mouse, Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012167

 
life technologies
LS13709681
50μg Each for $158.05

5012168

 
life technologies
LS13709685
500μg Each for $404.55
Description & Specifications

Specifications

Antigen CCL2 (MCP-1)
Clone 2H5
Host Species Armenian Hamster
Gene Alias MCP1, MCAF, CCL-2
Species Reactivity Human, Mouse, Rat
Applications ELISA Detection
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 2H5 monoclonal antibody reacts with mouse, rat, and human monocyte chemoattractant protein-1 (MCP-1), also known as CCL2 and MCAF.