missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CCL2 (MCP-1) Armenian Hamster anti-Human, Mouse, Rat, FITC, Clone: 2H5, eBioscience™

Armenian Hamster Monoclonal Antibody

$117.48 - $157.08

Specifications

Antigen CCL2 (MCP-1)
Clone 2H5
Host Species Armenian Hamster
Gene Alias MCP1, MCAF, CCL-2
Species Reactivity Human, Mouse, Rat
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010004 Life Technologies
LS11709641
25 Tests Each for $117.48
5010005 Life Technologies
LS11709681
50μg Each for $157.08
Specifications

Description

The 2H5 monoclonal antibody reacts with mouse, rat, and human monocyte chemoattractant protein-1 (MCP-1), also known as CCL2 and MCAF.
Description & Specifications

Specifications

CCL2 (MCP-1)
Armenian Hamster
Human, Mouse, Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
2H5
MCP1, MCAF, CCL-2
Flow Cytometry (Intracellular Staining)
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL; 5μL (0.5μg)/test