Please login to your online account to display your discounted pricing

CCG 63802, Tocris™

Inhibitor of regulator of G-protein signaling 4 (RGS4) protein

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material CCG 63802
Assay Percent Range >99%
Molecular Formula C26H18N4O2S
Formula Weight 450.51
Synonym (2E)-2-(1, 3-Benzothiazol-2-yl)-3-[9-methyl-2-(3-methylphenoxy)-4-oxo-4H-pyrido[1, 2-a]pyrimidin-3-yl]acrylonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

402810

 
r & d systems
RDS402810
10mg Each for $239.63

402850

 
r & d systems
RDS402850
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 450.51
Quantity 10mg
Chemical Name or Material CCG 63802
Assay Percent Range >99%
CAS 620112-78-9
Synonym (2E)-2-(1, 3-Benzothiazol-2-yl)-3-[9-methyl-2-(3-methylphenoxy)-4-oxo-4H-pyrido[1, 2-a]pyrimidin-3-yl]acrylonitrile
Molecular Formula C26H18N4O2S
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C