Please login to your online account to display your discounted pricing

β-CCB, Tocris


Benzodiazepine inverse agonist, putative endogenous ligand

Quantity 10mg

Manufacturer: r & d systems  040510

 View more versions of this product

Catalog No. 040510

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 268.31
CAS 84454-35-3
Assay Percent Range >99%
Molecular Formula C16H16N2O2
Chemical Name or Material β-CCB
Synonym Butyl β-carboline-3-carboxylate
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material β-CCB
Assay Percent Range >99%
Molecular Formula C16H16N2O2
Formula Weight 268.31
Synonym Butyl β-carboline-3-carboxylate
Accessories

Store at Room Temperature