Please login to your online account to display your discounted pricing

β-CCB, Tocris

Benzodiazepine inverse agonist, putative endogenous ligand

$133.98 - $567.83

Chemical Identifiers

Chemical Name or Material β-CCB
Assay Percent Range >99%
Molecular Formula C16H16N2O2
Formula Weight 268.31
Synonym Butyl β-carboline-3-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

040510

 
r & d systems
040510
10mg Each for $133.98

040550

 
r & d systems
040550
50mg Each for $567.83
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 268.31
Chemical Name or Material β-CCB
Assay Percent Range >99%
Molecular Formula C16H16N2O2
CAS 84454-35-3
Synonym Butyl β-carboline-3-carboxylate
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature