Please login to your online account to display your discounted pricing

CBZ-L-Serine methyleester, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material CBZ-L-Serine methyleester
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C12H15NO5
Formula Weight 253.26
Synonym N-Carbobenzoxy-L-serine methyleester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC434950100

  SDS
thermo fisher scientific
AC434950100
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC434950010

  SDS
thermo fisher scientific
AC434950010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Viscous Liquid or Solid
Assay ≥94.0%
Formula Weight 253.26
Quantity 10g
Color Pale yellow
Specific Rotation -12.5° to -14.4° (20°C, 589nm) (c-10, methanol)
Chemical Name or Material CBZ-L-Serine methyleester
Assay Percent Range 95%
Molecular Formula C12H15NO5
CAS 1676-81-9
Infrared Spectrum Authentic
Synonym N-Carbobenzoxy-L-serine methyleester