missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CBZ-L-Serine methyl ester, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC434950100

 View more versions of this product

Catalog No. AC434950100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

CBZ-L-Serine methyl ester
Authentic
-13.5
Yellow
1676-81-9
C12H15NO5
CINAUOAOVQPWIB-JTQLQIEISA-N
methyl (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoate
6999535
Viscous Liquid or Solid
113°C
Glass Bottle
170°C (0.1mmHg)
10g
94% min. (HPLC)
cbz-l-serine methyl ester, cbz-ser-ome, d-n-cbz-serine methyl ester, n-carbobenzoxy-l-serine methyl ester, n-carbobenzyloxy-l-serine methyl ester, n-cbz-l-serine methyl ester, n-z-l-serine methyl ester, s-methyl 2-benzyloxy carbonyl amino-3-hydroxypropanoate, z-l-serine methyl ester, z-ser-ome
COC(=O)C(CO)NC(=O)OCC1=CC=CC=C1
253.26
253.26
95%

Chemical Identifiers

1676-81-9
253.26
cbz-l-serine methyl ester, cbz-ser-ome, d-n-cbz-serine methyl ester, n-carbobenzoxy-l-serine methyl ester, n-carbobenzyloxy-l-serine methyl ester, n-cbz-l-serine methyl ester, n-z-l-serine methyl ester, s-methyl 2-benzyloxy carbonyl amino-3-hydroxypropanoate, z-l-serine methyl ester, z-ser-ome
methyl (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoate
C12H15NO5
CINAUOAOVQPWIB-JTQLQIEISA-N
6999535
COC(=O)C(CO)NC(=O)OCC1=CC=CC=C1
SDS Documents