CBS Scientific Gradient Markers
Linear with side outlets.
Supplier: CBS Scientific GM100
Description
- Gradient flow is controlled by a PTFE-coated valve centered between the two chambers
- The outflow channel is fitted with a male luer adapter which can be used with any needle or luer valve
Specifications
Electrophoresis | |
Gradient Marker | |
1.69 oz. | |
50 mL |