missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

cBRIDP(regR), TCI America™

Manufacturer:  TCI America C2979200MG

 View more versions of this product

Catalog No. C2979200MG



Specifications

Specifications

cBRIDP(regR)
91°C
742103-27-1
MFCD09753012
QMLPJDVGNRHGJQ-UHFFFAOYSA-N
ditert-butyl-(1-methyl-2,2-diphenylcyclopropyl)phosphane
21909131
Crystal-Powder at 20°C
White-Yellow
200mg
C24H33P
Di-tert-butyl(1-methyl-2,2-diphenylcyclopropyl)phosphine
CC1(CC1(C2=CC=CC=C2)C3=CC=CC=C3)P(C(C)(C)C)C(C)(C)C
352.502
352.50
≥97.0% (HPLC)

Chemical Identifiers

742103-27-1
352.502
QMLPJDVGNRHGJQ-UHFFFAOYSA-N
21909131
CC1(CC1(C2=CC=CC=C2)C3=CC=CC=C3)P(C(C)(C)C)C(C)(C)C
C24H33P
MFCD09753012
Di-tert-butyl(1-methyl-2,2-diphenylcyclopropyl)phosphine
ditert-butyl-(1-methyl-2,2-diphenylcyclopropyl)phosphane
Safety and Handling

Safety and Handling

TSCA : No

Documents