Please login to your online account to display your discounted pricing

CBiPES hydrochloride, Tocris™

Positive allosteric modulator of mGlu2

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material CBiPES hydrochloride
Assay Percent Range >99%
Molecular Formula C21H19N3O2S·HCl
Formula Weight 413.92
Synonym N-(4'-Cyano-[1, 1'-biphenyl]-3-yl-N-(3-pyridinylmethyl)-ethanesulfonamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

394950

 
r & d systems
RDS394950
50mg Each for $915.10

394910

 
r & d systems
RDS394910
10mg Each for $218.74
Description & Specifications

Specifications

Formula Weight 413.92
Quantity 50mg
Chemical Name or Material CBiPES hydrochloride
Assay Percent Range >99%
Molecular Formula C21H19N3O2S·HCl
CAS 856702-40-4
Synonym N-(4'-Cyano-[1, 1'-biphenyl]-3-yl-N-(3-pyridinylmethyl)-ethanesulfonamide hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature