Please login to your online account to display your discounted pricing

Alfa Aesar™ Cationic Acid Labile Surfactant II (CALS II), Certified Mass Spec Grade


Quantity 5mg

Manufacturer: alfa aesar™  J67525LB0

 View more versions of this product

Catalog No. AAJ67525LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Lyophilized Powder
Purity Grade Certified Mass Spec Grade
Odor Odorless
Quantity 5mg
Color White
Storage Note 1 -30° to -10°C
Chemical Name or Material Cationic Acid Labile Surfactant II (CALS II)

Specifications

Chemical Name or Material Cationic Acid Labile Surfactant II (CALS II)
Purity Grade Certified Mass Spec Grade
Accessories

-30° to -10°C

SDS & Safety Documents