Please login to your online account to display your discounted pricing

Alfa Aesar™ Cationic Acid Labile Surfactant I (CALS I), Certified Mass Spec Grade


Quantity 5mg

Manufacturer: alfa aesar™  J67743LB0

 View more versions of this product

Catalog No. AAJ67743LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Cationic Acid Labile Surfactant I (CALS I)
Physical Form Lyophilized Powder
Storage Note 1 -30° to -10°C
Purity Grade Certified Mass Spec Grade
Color White
Quantity 5mg
Odor Odorless

Specifications

Chemical Name or Material Cationic Acid Labile Surfactant I (CALS I)
Purity Grade Certified Mass Spec Grade
Accessories

-30° to -10°C

SDS & Safety Documents