Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Cathepsin G Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry procedures with Thermo Scientific™ Cathepsin G Ab-1, Mouse Monoclonal Antibody.

$241.86 - $1,156.33

Specifications

Antigen Cathepsin G Ab-1
Clone 19C3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1835R7

 
thermo scientific™ lab vision™
MS1835R7
7mL Each for $673.04

MS1835S

 
thermo scientific™ lab vision™
MS1835S
1mL Each for $1,156.33

MS1835S0

 
thermo scientific™ lab vision™
MS1835S0
100µL Each for $241.86

MS1835S1

 
thermo scientific™ lab vision™
MS1835S1
500µL Each for $808.82
Description & Specifications

Specifications

Antigen Cathepsin G Ab-1
Clone 19C3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Apoptosis
Type Primary
Immunogen Recombinant protein corresponding to 124aa from N terminus of human cathepsin G
Isotype IgG1

Obtain accurate, reproducible results in immunohistochemistry procedures with Thermo Scientific™ Cathepsin G Ab-1, Mouse Monoclonal Antibody.

Cathepsin G is a serine protease found in azurophilic granules of neutrophils involved in phagocytic lysis of engulfed forein particles. It is also involved in NK cell cytotoxicity.

Host Species: Mouse

Clone: 19C3

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: N-terminal 124aa

Immunogen: Recombinant protein corresponding to 124aa from N-terminus of human cathepsin G

Molecular Weight: 29 kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO