Please login to your online account to display your discounted pricing

Cathepsin B Inhibitor IV

Specifications

CAS 147859-80-1
Formula Weight 397.46g/mol
Synonym CA-074 ME; (L-3-trans-(Propylcarbamyl)oxirane-2-carbonyl)-L-isoleucyl-L-proline methyl ester
Storage Requirements −30° to −10°C
Format Powder
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64737MCR

 
alfa aesar™
J64737-MCR
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

CAS 147859-80-1
Formula Weight 397.46g/mol
Synonym CA-074 ME; (L-3-trans-(Propylcarbamyl)oxirane-2-carbonyl)-L-isoleucyl-L-proline methyl ester
Storage Requirements −30° to −10°C
Format Powder
For Use With (Application) A cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B.
Description White
Solubility Soluble in DMSO

A cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B.