Please login to your online account to display your discounted pricing

Cathepsin B Inhibitor III

An inhibitor of Cathepsin B

$435.46 - $435.46

Specifications

CAS 134448-10-5
Formula Weight 383.44g/mol
Synonym CA-074; [L-3-trans-(Propylcarbamoyl)oxirane-2-carbonyl]-L-isoleucyl-L-proline
Storage Requirements −30° to −10°C
Format Powder
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65347LB0

 
alfa aesar™
J65347LB0
Each for $435.46
Description & Specifications

Specifications

CAS 134448-10-5
Formula Weight 383.44g/mol
Synonym CA-074; [L-3-trans-(Propylcarbamoyl)oxirane-2-carbonyl]-L-isoleucyl-L-proline
Storage Requirements −30° to −10°C
Format Powder
For Use With (Application) An inhibitor of Cathepsin B
Description White
Solubility Soluble in methanol at 1mg/ml; Also soluble in DMSO or ethanol