missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Catenin, β Ab-2, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

$280.90 - $1,386.45

Specifications

Antigen Catenin, beta Ab-2
Applications Immunoprecipitation,Western Blotting,Immunoprecipitation,Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Chicken,Human,Mouse,Rat,Chicken,Human,Mouse,Rat,Chicken,Human,Mouse,Rat,Chicken,Human,Mouse,Rat
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
RB1491P Thermo Scientific™ Lab Vision™
RB1491P
Purified Ab with BSA and Azide 1mL Each for $1,286.12
RB1491P0 Thermo Scientific™ Lab Vision™
RB1491P0
Purified Ab with BSA and Azide 100μL Each for $280.90
RB1491P1 Thermo Scientific™ Lab Vision™
RB1491P1
Purified Ab with BSA and Azide 500μL Each for $778.33
RB1491P1ABX Thermo Scientific™ Lab Vision™
RB1491P1ABX
Ab without BSA/Azide (Frozen) 500μL Each for $859.08
RB1491PABX Thermo Scientific™ Lab Vision™
RB1491PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,386.45
RB1491R7 Thermo Scientific™ Lab Vision™
RB1491R7
Ready-to-use for IHC (UVLP) 7mL Each for $672.41
Specifications

Description

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Thermo Scientific™ Catenin, β Ab-2, Rabbit Polyclonal Antibody.

The catenins (α, Β and γ) are ubiquitously expressed cytoplasmic proteins associated with E-cadherin at cellular junctions. Β-catenin also binds to N-cadherin and co-immunoprecipitates with APC. Cadherin/catenin complexes are linked to the cytoskeleton via a direct association between α-actinin and α-catenin. Increased tyrosine phosphorylation can disrupt catenin-cadherin complexes, influencing cellular adhesion.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Chicken and Xenopus. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of the β-catenin protein

Molecular Weight: 92 kDa

Positive Control: MCF-7 cells, Breast CA

Cellular Localization: Cytoplasmic and cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Catenin, beta Ab-2
Polyclonal
Chicken,Human,Mouse,Rat,Chicken,Human,Mouse,Rat,Chicken,Human,Mouse,Rat,Chicken,Human,Mouse,Rat
RUO
Primary
IgG
Immunoprecipitation,Western Blotting,Immunoprecipitation,Western Blotting
Unlabeled
Rabbit
Cell Adhesion
A synthetic peptide from the C-terminus of the Beta-catenin protein