Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Catenin, β Ab-2, Rabbit Polyclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  RB1491P1ABX

 View more versions of this product

Catalog No. RB1491P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Catenin, beta Ab-2
Applications Immunoprecipitation
Applications Western Blotting
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Rabbit
Immunogen A synthetic peptide from the C-terminus of the Beta-catenin protein
Isotype IgG
Quantity 500μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Type Primary
Monoclonal or Polyclonal Polyclonal
Target Species Chicken
Target Species Human
Target Species Murine
Target Species Rat

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Thermo Scientific™ Catenin, β Ab-2, Rabbit Polyclonal Antibody.

The catenins (α, Β and γ) are ubiquitously expressed cytoplasmic proteins associated with E-cadherin at cellular junctions. Β-catenin also binds to N-cadherin and co-immunoprecipitates with APC. Cadherin/catenin complexes are linked to the cytoskeleton via a direct association between α-actinin and α-catenin. Increased tyrosine phosphorylation can disrupt catenin-cadherin complexes, influencing cellular adhesion.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, Rat, Chicken and Xenopus. Others not tested.

Epitope: C-terminal

Immunogen: A synthetic peptide from the C-terminus of the β-catenin protein

Molecular Weight: 92 kDa

Positive Control: MCF-7 cells, Breast CA

Cellular Localization: Cytoplasmic and cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO